(1-Ethyl-3,5-dimethyl-1H-pyrazol-4-yl)methanol

Supplier: Matrix Scientific

027641-500MG
101824-580EA 290.77 USD
101824-580
(1-Ethyl-3,5-dimethyl-1H-pyrazol-4-yl)methanol
(1-Ethyl-3,5-dimethyl-1H-pyrazol-4-yl)methanol

Hazard: Irritant

Formula: C₈H₁₄N₂O
MDL Number: MFCD00462165
CAS Number: 90152-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR