2-(6-Fluoro-1H-benzimidazol-2-yl)ethanamine hydrochloride

Supplier: Matrix Scientific

027638-500MG
101824-574EA 387.69 USD
101824-574
2-(6-Fluoro-1H-benzimidazol-2-yl)ethanamine hydrochloride
2-(6-Fluoro-1H-benzimidazol-2-yl)ethanamine hydrochloride

Hazard: Irritant

Formula: C₉H₁₁ClFN₃
MDL Number: MFCD11841214
CAS Number: 887405-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR