N-Methyl-N-[2-(4-methylpiperidin-1-yl)ethyl]amine

Supplier: Matrix Scientific

027632-500MG
101824-562EA 387.69 USD
101824-562
N-Methyl-N-[2-(4-methylpiperidin-1-yl)ethyl]amine
N-Methyl-N-[2-(4-methylpiperidin-1-yl)ethyl]amine

Hazard: Irritant

Formula: C₉H₂₀N₂
MDL Number: MFCD08691570
CAS Number: 915924-43-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR