N-[2-(4-Methylpiperidin-1-yl)ethyl]propan-2-amine

Supplier: Matrix Scientific

027630-500MG
101824-558EA 387.69 USD
101824-558
N-[2-(4-Methylpiperidin-1-yl)ethyl]propan-2-amine
N-[2-(4-Methylpiperidin-1-yl)ethyl]propan-2-amine

Hazard: Irritant

Formula: C₁₁H₂₄N₂
MDL Number: MFCD08691568
CAS Number: 915924-65-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR