N-(2-Azepan-1-ylethyl)-N-isopropylamine

Supplier: Matrix Scientific

027629-500MG
101824-556EA 387.69 USD
101824-556
N-(2-Azepan-1-ylethyl)-N-isopropylamine
N-(2-Azepan-1-ylethyl)-N-isopropylamine

Hazard: Irritant

Formula: C₁₁H₂₄N₂
MDL Number: MFCD08691567
CAS Number: 532407-05-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR