5,7-Dimethyl[1,2,4]triazolo[4,3-a]pyrimidine-3-carboxylic acid

Supplier: Matrix Scientific

Synonyms: 5,7-Dimethyl-[1,2,4]triazolo[4,3-a]pyrimidine-3-carboxylic acid

027627-500MG
101824-552EA 640 USD
101824-552
5,7-Dimethyl[1,2,4]triazolo[4,3-a]pyrimidine-3-carboxylic acid
5,7-Dimethyl[1,2,4]triazolo[4,3-a]pyrimidine-3-carboxylic acid

Hazard: Irritant

Formula: C₈H₈N₄O₂
MW: 192.18 g/mol
MDL Number: MFCD05668696
CAS Number: 842972-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR