3-(3,4-Dihydroisoquinolin-2(1H)-yl)propan-1-ol

Supplier: Matrix Scientific

027624-500MG
101824-546EA 233.85 USD
101824-546
3-(3,4-Dihydroisoquinolin-2(1H)-yl)propan-1-ol
3-(3,4-Dihydroisoquinolin-2(1H)-yl)propan-1-ol

Hazard: Irritant

Formula: C₁₂H₁₇NO
MDL Number: MFCD08691565
CAS Number: 86368-07-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR