N-Ethyl-N-[2-(5-methoxy-1H-benzimidazol-2-yl)ethyl]amine

Supplier: Matrix Scientific

027610-500MG
101824-518EA 387.69 USD
101824-518
N-Ethyl-N-[2-(5-methoxy-1H-benzimidazol-2-yl)ethyl]amine
N-Ethyl-N-[2-(5-methoxy-1H-benzimidazol-2-yl)ethyl]amine

Hazard: Irritant

Formula: C₁₂H₁₇N₃O
MW: 219.29 g/mol
MDL Number: MFCD08691497
CAS Number: 915924-04-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR