N-(2-Methoxyethyl)-N-[(1,3,5-trimethyl-1H-pyrazol-4-yl)methyl]amine

Supplier: Matrix Scientific

027606-500MG
101824-510EA 387.69 USD
101824-510
N-(2-Methoxyethyl)-N-[(1,3,5-trimethyl-1H-pyrazol-4-yl)methyl]amine
N-(2-Methoxyethyl)-N-[(1,3,5-trimethyl-1H-pyrazol-4-yl)methyl]amine

Hazard: Irritant

Formula: C₁₀H₁₉N₃O
MDL Number: MFCD08691493
CAS Number: 1015845-58-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR