N-Isobutyl-N-[(1,3,5-trimethyl-1H-pyrazol-4-yl)methyl]amine

Supplier: Matrix Scientific

027605-500MG
101824-508EA 387.69 USD
101824-508
N-Isobutyl-N-[(1,3,5-trimethyl-1H-pyrazol-4-yl)methyl]amine
N-Isobutyl-N-[(1,3,5-trimethyl-1H-pyrazol-4-yl)methyl]amine

Hazard: Irritant

Formula: C₁₁H₂₁N₃
MW: 195.31 g/mol
MDL Number: MFCD08691491
CAS Number: 1152873-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR