[(4-Methyl-1H-benzimidazol-2-yl)methoxy]acetic acid

Supplier: Matrix Scientific

027604-500MG
101824-506EA 290.77 USD
101824-506
[(4-Methyl-1H-benzimidazol-2-yl)methoxy]acetic acid
[(4-Methyl-1H-benzimidazol-2-yl)methoxy]acetic acid

Hazard: Irritant

Formula: C₁₁H₁₂N₂O₃
MDL Number: MFCD08691490
CAS Number: 915924-02-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR