2-(5,6-Dimethyl-1H-benzimidazol-2-yl)ethanamine

Supplier: Matrix Scientific

027596-500MG
101824-490EA 584.62 USD
101824-490
2-(5,6-Dimethyl-1H-benzimidazol-2-yl)ethanamine
2-(5,6-Dimethyl-1H-benzimidazol-2-yl)ethanamine

Hazard: Irritant

Formula: C₁₁H₁₅N₃
MDL Number: MFCD08691441
CAS Number: 91337-46-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR