2-(2-Methyl-1H-benzo[d]imidazol-1-yl)propanoic acid

Supplier: Matrix Scientific

027594-500MG
101824-486EA 290.77 USD
101824-486
2-(2-Methyl-1H-benzo[d]imidazol-1-yl)propanoic acid
2-(2-Methyl-1H-benzo[d]imidazol-1-yl)propanoic acid

Hazard: Irritant

Formula: C₁₁H₁₂N₂O₂
MDL Number: MFCD03161170
CAS Number: 753489-92-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR