(2-Cyclopropyl-1H-benzimidazol-1-yl)acetic acid

Supplier: Matrix Scientific

027593-500MG
101824-484EA 387.69 USD
101824-484
(2-Cyclopropyl-1H-benzimidazol-1-yl)acetic acid
(2-Cyclopropyl-1H-benzimidazol-1-yl)acetic acid

Hazard: Irritant

Formula: C₁₂H₁₂N₂O₂
MDL Number: MFCD08691428
CAS Number: 97968-85-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR