(2-Isopropyl-1H-benzimidazol-1-yl)acetic acid

Supplier: Matrix Scientific

027592-500MG
101824-482EA 461.54 USD
101824-482
(2-Isopropyl-1H-benzimidazol-1-yl)acetic acid
(2-Isopropyl-1H-benzimidazol-1-yl)acetic acid

Hazard: Irritant

Formula: C₁₂H₁₄N₂O₂
MDL Number: MFCD05239272
CAS Number: 797812-91-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR