5-Bromo-7-methyl-2,3-dihydrofuro[2,3-f][1,4]benzodioxine-8-carboxylic acid

Supplier: Matrix Scientific

027586-500MG
101824-472EA 387.69 USD
101824-472
5-Bromo-7-methyl-2,3-dihydrofuro[2,3-f][1,4]benzodioxine-8-carboxylic acid
5-Bromo-7-methyl-2,3-dihydrofuro[2,3-f][1,4]benzodioxine-8-carboxylic acid

Hazard: Irritant

Formula: C₁₂H₉BrO₅
MW: 313.11 g/mol
MDL Number: MFCD00514040
CAS Number: 933682-36-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR