N-(1H-Benzimidazol-2-ylmethyl)-N-phenylamine
Supplier: Matrix Scientific
027585-500MG
101824-470EA
387.69
USD
101824-470
N-(1H-Benzimidazol-2-ylmethyl)-N-phenylamine
N-(1H-Benzimidazol-2-ylmethyl)-N-phenylamine
Hazard: Irritant
Formula:
C₁₄H₁₃N₃ |
MDL Number:
MFCD06799591 CAS Number: 5805-59-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...