(4-Aminopiperidin-1-yl)acetic acid

Supplier: Matrix Scientific

027579-500MG
101824-458EA 387.69 USD
101824-458
(4-Aminopiperidin-1-yl)acetic acid
(4-Aminopiperidin-1-yl)acetic acid

Hazard: Irritant

Formula: C₇H₁₄N₂O₂
MDL Number: MFCD08848201
CAS Number: 805179-70-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR