4-[2-(1H-Imidazol-1-yl)ethyl]piperidine

Supplier: Matrix Scientific

027578-500MG
101824-456EA 323.08 USD
101824-456
4-[2-(1H-Imidazol-1-yl)ethyl]piperidine
4-[2-(1H-Imidazol-1-yl)ethyl]piperidine

Hazard: Irritant

Formula: C₁₀H₁₇N₃
MDL Number: MFCD08848193
CAS Number: 90747-20-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR