(5-Methyl-1H-tetrazol-1-yl)(phenyl)acetic acid

Supplier: Matrix Scientific

027567-500MG
101824-432EA 387.69 USD
101824-432
(5-Methyl-1H-tetrazol-1-yl)(phenyl)acetic acid
(5-Methyl-1H-tetrazol-1-yl)(phenyl)acetic acid

Hazard: Irritant

Formula: C₁₀H₁₀N₄O₂
MDL Number: MFCD07643231
CAS Number: 876716-32-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR