1-(4-Aminophenyl)-1H-tetrazole
Supplier: Matrix Scientific
027559-1G
101824-416EA
372.31
USD
101824-416
1-(4-Aminophenyl)-1H-tetrazole
1-(4-Aminophenyl)-1H-tetrazole
Hazard: Irritant
Formula:
C₇H₇N₅ |
MDL Number:
MFCD02746184 CAS Number: 14213-13-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...