4-(3-Aminopropyl)-1,3-thiazol-2-amine
Supplier: Matrix Scientific
027551-500MG
101824-400EA
387.69
USD
101824-400
4-(3-Aminopropyl)-1,3-thiazol-2-amine
4-(3-Aminopropyl)-1,3-thiazol-2-amine
Hazard: Irritant
Formula:
C₆H₁₁N₃S |
MDL Number:
MFCD08059800 CAS Number: 136604-78-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...