1',5'-Dimethyl-1'H,2H-3,4'-bipyrazol-5-amine

Supplier: Matrix Scientific

027541-500MG
101824-382EA 3076.92 USD
101824-382
1',5'-Dimethyl-1'H,2H-3,4'-bipyrazol-5-amine
1',5'-Dimethyl-1'H,2H-3,4'-bipyrazol-5-amine

Hazard: Irritant

Formula: C₈H₁₁N₅
MDL Number: MFCD08685886
CAS Number: 1025026-56-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR