4-(Difluoromethoxy)-3-methoxybenzoic acid
Supplier: Matrix Scientific
027539-500MG
101824-376EA
193.85
USD
101824-376
4-(Difluoromethoxy)-3-methoxybenzoic acid
4-(Difluoromethoxy)-3-methoxybenzoic acid
Hazard: Irritant
Formula:
C₉H₈F₂O₄ |
MDL Number:
MFCD03964657 CAS Number: 162401-59-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...