2,7-Dimethylquinoline-4-carboxylic acid

Supplier: Matrix Scientific

027532-500MG
101824-362EA 193.85 USD
101824-362
2,7-Dimethylquinoline-4-carboxylic acid
2,7-Dimethylquinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₁NO₂
MDL Number: MFCD08681035
CAS Number: 892674-22-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR