(6-Fluoro-1-methyl-2-oxo-2,3-dihydro-1H-indol-3-yl)acetic acid

Supplier: Matrix Scientific

027531-500MG
101824-360EA 495.38 USD
101824-360
(6-Fluoro-1-methyl-2-oxo-2,3-dihydro-1H-indol-3-yl)acetic acid
(6-Fluoro-1-methyl-2-oxo-2,3-dihydro-1H-indol-3-yl)acetic acid

Hazard: Irritant

Formula: C₁₁H₁₀FNO₃
MDL Number: MFCD09971852
CAS Number: 933752-43-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR