5-Methoxy-2-oxoindoline-3-carbaldehyde

Supplier: Matrix Scientific

027529-500MG
101824-356EA 387.69 USD
101824-356
5-Methoxy-2-oxoindoline-3-carbaldehyde
5-Methoxy-2-oxoindoline-3-carbaldehyde

Hazard: Irritant

Formula: C₁₀H₉NO₃
MDL Number: MFCD08691731
CAS Number: 52508-88-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR