3-Amino-6-methoxy-1,3-dihydro-2H-indol-2-one

Supplier: Matrix Scientific

027526-500MG
101824-350EA 387.69 USD
101824-350
3-Amino-6-methoxy-1,3-dihydro-2H-indol-2-one
3-Amino-6-methoxy-1,3-dihydro-2H-indol-2-one

Hazard: Irritant

Formula: C₉H₁₀N₂O₂
MDL Number: MFCD08691527
CAS Number: 1105682-79-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR