3-Amino-6-fluoro-1,3-dihydro-2H-indol-2-one hydrochloride
Supplier: Matrix Scientific
027525-500MG
101824-348EA
387.69
USD
101824-348
3-Amino-6-fluoro-1,3-dihydro-2H-indol-2-one hydrochloride
3-Amino-6-fluoro-1,3-dihydro-2H-indol-2-one hydrochloride
Hazard: Irritant
Formula:
C₈H₈ClFN₂O |
MDL Number:
MFCD11841258 CAS Number: 1214059-84-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...