3,6-Dimethyl-2-thioxohexahydropyrimidine-4-carboxylic acid
Supplier: Matrix Scientific
027520-500MG
101824-338EA
387.69
USD
101824-338
3,6-Dimethyl-2-thioxohexahydropyrimidine-4-carboxylic acid
3,6-Dimethyl-2-thioxohexahydropyrimidine-4-carboxylic acid
Hazard: Irritant
Formula:
C₇H₁₂N₂O₂S |
MDL Number:
MFCD09971848 CAS Number: 1103826-96-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...