Methyl (1-methyl-5-oxo-4,5-dihydro-1H-pyrrol-2-yl)acetate

Supplier: Matrix Scientific

027518-500MG
101824-334EA 387.69 USD
101824-334
Methyl (1-methyl-5-oxo-4,5-dihydro-1H-pyrrol-2-yl)acetate
Methyl (1-methyl-5-oxo-4,5-dihydro-1H-pyrrol-2-yl)acetate

Hazard: Irritant

Formula: C₈H₁₁NO₃
MDL Number: MFCD09971847
CAS Number: 887405-61-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR