5-Chloro-2-oxoindoline-3-carbaldehyde

Supplier: Matrix Scientific

027515-500MG
101824-328EA 340 USD
101824-328
5-Chloro-2-oxoindoline-3-carbaldehyde
5-Chloro-2-oxoindoline-3-carbaldehyde

Hazard: Irritant

Formula: C₉H₆ClNO₂
MDL Number: MFCD09971844
CAS Number: 52508-86-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR