3-Amino-4-methyl-1,3-dihydro-2H-indol-2-one hydrochloride

Supplier: Matrix Scientific

027513-500MG
101824-322EA 300 USD
101824-322
3-Amino-4-methyl-1,3-dihydro-2H-indol-2-one hydrochloride
3-Amino-4-methyl-1,3-dihydro-2H-indol-2-one hydrochloride

Hazard: Irritant

Formula: C₉H₁₁ClN₂O
MDL Number: MFCD11502899
CAS Number: 1214103-58-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR