3-Chloro-6-isopropoxypyridazine

Supplier: Matrix Scientific

027511-500MG
101824-318EA 387.69 USD
101824-318
3-Chloro-6-isopropoxypyridazine
3-Chloro-6-isopropoxypyridazine

Hazard: Irritant

Formula: C₇H₉ClN₂O
MDL Number: MFCD00270293
CAS Number: 3184-71-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR