4-Bromo-2,5-dimethoxybenzaldehyde

Supplier: Matrix Scientific

027502-500MG
101824-302EA 289.8 USD
101824-302
4-Bromo-2,5-dimethoxybenzaldehyde
4-Bromo-2,5-dimethoxybenzaldehyde

Hazard: Irritant

Formula: C₉H₉BrO₃
MW: 245.08 g/mol
MDL Number: MFCD00156964
CAS Number: 31558-41-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR