3-(3,5-Dimethylisoxazol-4-yl)propan-1-amine

Supplier: Matrix Scientific

027499-500MG
101824-296EA 495.38 USD
101824-296
3-(3,5-Dimethylisoxazol-4-yl)propan-1-amine
3-(3,5-Dimethylisoxazol-4-yl)propan-1-amine

Hazard: Irritant

Formula: C₈H₁₄N₂O
MDL Number: MFCD06213632
CAS Number: 771572-98-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR