1-Hydroxy-4-phenyl-1H-1,2,3-triazole-5-carboxylic acid

Supplier: Matrix Scientific

027494-500MG
101824-288EA 461.54 USD
101824-288
1-Hydroxy-4-phenyl-1H-1,2,3-triazole-5-carboxylic acid
1-Hydroxy-4-phenyl-1H-1,2,3-triazole-5-carboxylic acid

Hazard: Irritant

Formula: C₉H₇N₃O₃
MDL Number: MFCD09971839
CAS Number: 887405-55-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR