1-[(4-Aminophenyl)sulfonyl]piperidine-4-carboxylic acid

Supplier: Matrix Scientific

027493-500MG
101824-286EA 387.69 USD
101824-286
1-[(4-Aminophenyl)sulfonyl]piperidine-4-carboxylic acid
1-[(4-Aminophenyl)sulfonyl]piperidine-4-carboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₆N₂O₄S
MDL Number: MFCD03281552
CAS Number: 436092-84-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR