5-(Aminomethyl)[1,2,4]triazolo[1,5-a]pyrimidin-7(4H)-one

Supplier: Matrix Scientific

027489-500MG
101824-278EA 387.69 USD
101824-278
5-(Aminomethyl)[1,2,4]triazolo[1,5-a]pyrimidin-7(4H)-one
5-(Aminomethyl)[1,2,4]triazolo[1,5-a]pyrimidin-7(4H)-one

Hazard: Irritant

Formula: C₆H₇N₅O
MDL Number: MFCD09971836
CAS Number: 887405-49-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR