1-Isothiocyanato-3-(trifluoromethoxy)benzene

Supplier: Matrix Scientific

027488-500MG
101824-276EA 387.69 USD
101824-276
1-Isothiocyanato-3-(trifluoromethoxy)benzene
1-Isothiocyanato-3-(trifluoromethoxy)benzene

Hazard: Irritant

Formula: C₈H₄F₃NOS
MDL Number: MFCD07596437
CAS Number: 471937-78-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR