1-(5-Amino-2-methoxybenzyl)pyrrolidin-2-one hydrochloride

Supplier: Matrix Scientific

027483-500MG
101824-268EA 384.62 USD
101824-268
1-(5-Amino-2-methoxybenzyl)pyrrolidin-2-one hydrochloride
1-(5-Amino-2-methoxybenzyl)pyrrolidin-2-one hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇ClN₂O₂
MDL Number: MFCD11506409
CAS Number: 1185294-41-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR