3-(1H-Imidazol-1-yl)butanoic acid
Supplier: Matrix Scientific
027482-500MG
101824-266EA
387.69
USD
101824-266
3-(1H-Imidazol-1-yl)butanoic acid
3-(1H-Imidazol-1-yl)butanoic acid
Hazard: Irritant
Formula:
C₇H₁₀N₂O₂ |
MDL Number:
MFCD00749150 CAS Number: 98009-60-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...