N-1-Adamantyl-N-methylamine

Supplier: Matrix Scientific

Synonyms: N-Methyladamantan-1-amine

027476-500MG
101824-254EA 198.46 USD
101824-254
N-1-Adamantyl-N-methylamine
N-1-Adamantyl-N-methylamine

Hazard: Irritant

Formula: C₁₁H₁₉N
MW: 165.28 g/mol
MDL Number: MFCD01674017
CAS Number: 3717-38-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR