5-Hydroxy-2-adamantanone ≥97%
Supplier: Matrix Scientific
Synonyms:
5-Hydroxyadamantan-2-one
027475-1G
101824-250EA
15.38
USD
101824-250
5-Hydroxy-2-adamantanone ≥97%
5-Hydroxy-2-adamantanone
Hazard: Irritant
Formula:
C₁₀H₁₄O₂ MW: 166.22 g/mol |
MDL Number:
MFCD00192211 CAS Number: 20098-14-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...