Di-adamantan-2-ylamine

Supplier: Matrix Scientific

027474-500MG
101824-248EA 387.69 USD
101824-248
Di-adamantan-2-ylamine
Di-adamantan-2-ylamine

Hazard: Irritant

Formula: C₂₀H₃₁N
MDL Number: MFCD02641388
CAS Number: 39234-40-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR