[3-(3,4-Dimethylphenyl)-1-adamantyl]methylamine

Supplier: Matrix Scientific

027472-500MG
101824-244EA 387.69 USD
101824-244
[3-(3,4-Dimethylphenyl)-1-adamantyl]methylamine
[3-(3,4-Dimethylphenyl)-1-adamantyl]methylamine

Hazard: Irritant

Formula: C₁₉H₂₇N
MDL Number: MFCD01821206
CAS Number: 400749-85-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR