N-[(1-Isobutylpiperidin-4-yl)methyl]-N-methylamine

Supplier: Matrix Scientific

027467-500MG
101824-234EA 387.69 USD
101824-234
N-[(1-Isobutylpiperidin-4-yl)methyl]-N-methylamine
N-[(1-Isobutylpiperidin-4-yl)methyl]-N-methylamine

Hazard: Irritant

Formula: C₁₁H₂₄N₂
MDL Number: MFCD08361797
CAS Number: 887405-46-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR