1-(1-Ethyl-5-methyl-1H-pyrazol-4-yl)ethanone

Supplier: Matrix Scientific

027466-500MG
101824-232EA 173.85 USD
101824-232
1-(1-Ethyl-5-methyl-1H-pyrazol-4-yl)ethanone
1-(1-Ethyl-5-methyl-1H-pyrazol-4-yl)ethanone

Hazard: Irritant

Formula: C₈H₁₂N₂O
MDL Number: MFCD03167209
CAS Number: 946658-63-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR