6-Hydrazino-2-phenyl-3H-imidazo[4,5-b]pyridine

Supplier: Matrix Scientific

027465-500MG
101824-230EA 387.69 USD
101824-230
6-Hydrazino-2-phenyl-3H-imidazo[4,5-b]pyridine
6-Hydrazino-2-phenyl-3H-imidazo[4,5-b]pyridine

Hazard: Irritant

Formula: C₁₂H₁₁N₅
MDL Number: MFCD09971831
CAS Number: 887405-44-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR