1,4-Dioxa-7-azaspiro[4.4]nonane

Supplier: Matrix Scientific

027463-500MG
101824-228EA 492.31 USD
101824-228
1,4-Dioxa-7-azaspiro[4.4]nonane
1,4-Dioxa-7-azaspiro[4.4]nonane

Hazard: Irritant

Formula: C₆H₁₁NO₂
MDL Number: MFCD09833560
CAS Number: 176-33-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR